Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ cis-Dichlorobis(pyridine)platinum(II), 99%

Click to view available options
Quantity:
0.25g
1g
14872-21-0
C10H10Cl2N2Pt
424.188
MFCD00058858
FCJXCLMZOWAKNQ-UHFFFAOYSA-L
dichloroplatinum; pyridine,platinum cis-dichlorobispyridine,platinum ii chloride; bis pyridine,dichlorobis pyridine platinum ii,cis-dichlorobis pyridine platinum ii,platinum, dichloro-, compd. with pyridine 1:2
498600
dichloroplatinum;pyridine
C1=CC=NC=C1.C1=CC=NC=C1.Cl[Pt]Cl
14872-21-0
C10H10Cl2N2Pt
424.188
MFCD00058858
FCJXCLMZOWAKNQ-UHFFFAOYSA-L
dichloroplatinum; pyridine,platinum cis-dichlorobispyridine,platinum ii chloride; bis pyridine,dichlorobis pyridine platinum ii,cis-dichlorobis pyridine platinum ii,platinum, dichloro-, compd. with pyridine 1:2
498600
dichloroplatinum;pyridine
C1=CC=NC=C1.C1=CC=NC=C1.Cl[Pt]Cl
Melting Point 282°C to 288°C
Quantity 1g
Formula Weight 424.19
Percent Purity 99%
Chemical Name or Material cis-Dichlorobis(pyridine)platinum(II)

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger