Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-Epoxysuccinic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
16533-72-5
C4H4O5
132.07
MFCD00191645
DCEMCPAKSGRHCN-XIXRPRMCSA-N
cis-epoxysuccinic acid,2r,3s-oxirane-2,3-dicarboxylic acid,cis-oxirane-2,3-dicarboxylic acid,2,3-oxiranedicarboxylic acid, 2r,3s-rel,2,3-oxiranedicarboxylic acid, 2r,3s-rel-9ci
2734802
(2S,3R)-oxirane-2,3-dicarboxylic acid
C1(C(O1)C(=O)O)C(=O)O
16533-72-5
C4H4O5
132.07
MFCD00191645
DCEMCPAKSGRHCN-XIXRPRMCSA-N
cis-epoxysuccinic acid,2r,3s-oxirane-2,3-dicarboxylic acid,cis-oxirane-2,3-dicarboxylic acid,2,3-oxiranedicarboxylic acid, 2r,3s-rel,2,3-oxiranedicarboxylic acid, 2r,3s-rel-9ci
2734802
(2S,3R)-oxirane-2,3-dicarboxylic acid
C1(C(O1)C(=O)O)C(=O)O
Melting Point 133°C to 135°C
Packaging Glass Bottle
Quantity 5g
Assay Percent Range 97%
Solubility Information Solubility in water: .. Other solubilities: soluble in organic solvents
Formula Weight 132.07
Percent Purity 97%
Chemical Name or Material cis-Epoxysuccinic acid

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

EINECSNumber : 240-604-3