Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
cis-Epoxysuccinic acid, 97%, ACROS Organics™
Click to view available options
Quantity:
5g
25g
Specifications
Melting Point | 133°C to 135°C |
Quantity | 25g |
Assay Percent Range | 97% |
Solubility Information | Solubility in water: .. Other solubilities: soluble in organic solvents |
Formula Weight | 132.07 |
Percent Purity | 97% |
Chemical Name or Material | cis-Epoxysuccinic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
EINECSNumber : 240-604-3