Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ cis-Stilbene, 97%

Click to view available options
Quantity:
1g
5g
25g
645-49-8
C14H12
180.25
MFCD00004788
PJANXHGTPQOBST-QXMHVHEDSA-N
cis-stilbene, z-stilbene, z-1,2-diphenylethene, cis-1,2-diphenylethylene, isostilbene, cis-diphenylethene, stilbene, z, z-2-phenylethenyl benzene, benzene, 1,1'-1,2-ethenediyl bis-, z, z-1,2-diphenylethylene
5356785
CHEBI:36008
(Z)-stilbene
C1=CC=C(C=C1)C=CC2=CC=CC=C2
645-49-8
C14H12
180.25
MFCD00004788
PJANXHGTPQOBST-QXMHVHEDSA-N
cis-stilbene, z-stilbene, z-1,2-diphenylethene, cis-1,2-diphenylethylene, isostilbene, cis-diphenylethene, stilbene, z, z-2-phenylethenyl benzene, benzene, 1,1'-1,2-ethenediyl bis-, z, z-1,2-diphenylethylene
5356785
CHEBI:36008
(Z)-stilbene
C1=CC=C(C=C1)C=CC2=CC=CC=C2
Applications
cis-Stilbene is used as a phosphor and a scintillator. It is mainly used in the produciton of dyes and optical brighteners.

Solubility
Immiscible with water. Miscible with ethanol.

Notes
Incompatible with oxidizing agents.
Sensitivity Light sensitive
Refractive Index 1.622
Density 1.04
Boiling Point 307°C
Flash Point >110°C (230°F)
Odor Weak
Quantity 1g
Melting Point 1°C to 2°C
Beilstein 1616739
Merck Index 14,8817
Solubility Information Immiscible with water. Miscible with ethanol.
Formula Weight 180.25
Percent Purity 97%
Chemical Name or Material cis-Stilbene
Show More Show Less

GHS H Statement
H319
Causes serious eye irritation.

GHS P Statement
P305+P351+P338
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.

Warning

EINECSNumber : 211-445-7

RTECSNumber : DA0513600

TSCA : No

Recommended Storage : Ambient temperatures