Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CISD1 Mouse anti-Human, Alexa Fluor™ 750, Clone: 1A8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP222567AF750

Catalog No. NP22267A70


Add to cart

Description

Description

CISD1 Monoclonal antibody specifically detects CISD1 in Human samples. It is validated for Western Blot,Flow Cytometry,ELISA.
Specifications

Specifications

CISD1
1A8
Western Blot, Flow Cytometry, ELISA
C10orf70, CDGSH iron sulfur domain 1, CDGSH iron sulfur domain-containing protein 1, CDGSH iron-sulfur domain-containing protein 1, chromosome 10 open reading frame 70, MDS029, MGC14684, mitoNEET, ZCD1, zinc finger CDGSH-type domain 1, zinc finger, CDGSH-type domain 1
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Human
ELISA, Flow Cytometry, Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
CISD1
Recombinant human CISD1 (32-108aa) purified from E. coli
0.1 mL
Primary
55847
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit