Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cisplatin, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
250 mg
1 g
25 g
Packaging:
Amber Glass Bottle
15663-27-1
Cl2H8N2Pt
302.06
OJMIKGOOWBVMEV-UHFFFAOYSA-N
cisplatin
N.N.Cl.Cl.[Pt]
15663-27-1
Cl2H8N2Pt
302.06
OJMIKGOOWBVMEV-UHFFFAOYSA-N
cisplatin
N.N.Cl.Cl.[Pt]

C1668, 15663-27-1, Cl2H6N2Pt

Spectrum™ Cisplatin, USP is part of the class of drugs known as platinum-containing anti-cancer drugs. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Quantity 250 mg
Packaging Amber Glass Bottle
CAS Max % 100%
UN Number UN3288
Percent Purity 98-102%