Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citalopram Hydrobromide, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
100 g
1 kg
Packaging:
Amber Glass Bottle
59729-32-7
C20H22BrFN2O
405.31
WIHMBLDNRMIGDW-UHFFFAOYNA-N
hydrogen 1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-2-benzofuran-5-carbonitrile bromide
[H+].[Br-].CN(C)CCCC1(OCC2=CC(=CC=C12)C#N)C1=CC=C(F)C=C1
59729-32-7
C20H22BrFN2O
405.31
WIHMBLDNRMIGDW-UHFFFAOYNA-N
hydrogen 1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-2-benzofuran-5-carbonitrile bromide
[H+].[Br-].CN(C)CCCC1(OCC2=CC(=CC=C12)C#N)C1=CC=C(F)C=C1

C2408, 59729-32-7, C20H21FN2O•HBr

Spectrum™ Citalopram Hydrobromide, USP is used as an antidepressant drug. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Heavy Metals 20 ppm
Ignition Residue 0.1%
Percent Purity 98-102%
Packaging Amber Glass Bottle
Specific Rotation -0.2° to +0.2°
Quantity 100 g