Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CITED1 Mouse anti-Human, Clone: OTI1C6, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526831

Catalog No. PIMA526831


Add to cart

Description

Description

This gene encodes a member of the CREB-binding protein/p300-interacting transactivator with Asp/Glu-rich C-terminal domain (CITED) family of proteins. The encoded protein, also known as melanocyte-specific gene 1, may function as a transcriptional coactivator and may play a role in pigmentation of melanocytes. Alternatively spliced transcript variants have been described.
Specifications

Specifications

CITED1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q99966
Cbp/p300-interacting transactivator; with Glu/Asp-rich carboxy-terminal domain; 1; MSG1
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4435
Western Blot
OTI1C6
Unconjugated
CITED1
Liquid
CITED1
Full length human recombit protein of CITED1 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit