Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CITED2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103000
Description
Antibody detects endogenous levels of total CITED2.
The protein encoded by this gene inhibits transactivation of HIF1A-induced genes by competing with binding of HIF1a to p300-CH1. Mutations in this gene are a cause of cardiac septal defects. Three transcript variants encoding the same protein have been found for this gene.
Specifications
CITED2 | |
Polyclonal | |
Unconjugated | |
CITED2 | |
AI835299; ASD8; Cbp/p300 interacting transactivator with Glu/Asp rich carboxy-terminal domain 2; cbp/p300-interacting transactivator 2; Cbp/p300-interacting transactivator, with Glu/Asp rich carboxy-terminal domain, 2; Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 2; Cited2; ER154-like; melanocyte specific gene 2; melanocyte-specific gene 1-related gene 1; Mrg1; MRG-1; MSG1-related gene 1; Msg2; MSG-related protein 1; P35srj; VSD2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
10370, 114490, 17684 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O35740, Q99967 | |
CITED2 | |
A synthesized peptide derived from human CITED2(Accession Q99967), corresponding to amino acid residues Y134-G184. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction