Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CITED4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568405
Description
CITED4 Polyclonal Antibody for IHC (P)
Cited4 is a member of The CITED family proteins that bind to CBP/p300 transcriptional integrators through their conserved C-terminal acidic domain and function as coactivators. It also interacts with isoforms of the TFAP2 transcription factor, coactivating TFAP2-dependent transcription.
Specifications
CITED4 | |
Polyclonal | |
Unconjugated | |
CITED4 | |
Cbp/p300 interacting transactivator with Glu/Asp rich carboxy-terminal domain 4; cbp/p300-interacting transactivator 4; Cbp/p300-interacting transactivator, with Glu/Asp rich carboxy-terminal domain, 4; Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 4; cited4; cited4 {ECO:0000312; Mrg2; MRG-2; MSG1-related protein 2; RGD:620113}; transcriptional co-activator 4 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
163732 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
2.3 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q96RK1 | |
CITED4 | |
Synthetic peptide corresponding to residues near the N terminal of human CITED4. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction