Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CITED4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568835
Description
This target displays homology in the following species: Mouse: 100%; Rat: 100%.
Cited4 is a member of The CITED family proteins that bind to CBP/p300 transcriptional integrators through their conserved C-terminal acidic domain and function as coactivators. It also interacts with isoforms of the TFAP2 transcription factor, coactivating TFAP2-dependent transcription.
Specifications
CITED4 | |
Polyclonal | |
Unconjugated | |
CITED4 | |
Cbp/p300 interacting transactivator with Glu/Asp rich carboxy-terminal domain 4; cbp/p300-interacting transactivator 4; Cbp/p300-interacting transactivator, with Glu/Asp rich carboxy-terminal domain, 4; Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 4; cited4; cited4 {ECO:0000312; Mrg2; MRG-2; MSG1-related protein 2; RGD:620113}; transcriptional co-activator 4 | |
Rabbit | |
Protein A | |
RUO | |
56222 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1.0 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9WUL8 | |
CITED4 | |
synthetic peptide directed towards the N terminal region of mouse CITED4. | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction