Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Citidine Deaminase Sheep anti-Human, Yeast, Polyclonal, Novus Biologicals™
Sheep Polyclonal Antibody
Specifications
Antigen | Citidine Deaminase |
---|---|
Concentration | 5.0 mg/mL |
Dilution | Western Blot 1:100-1:500, ELISA 1:500-1:2500, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10066413
|
Novus Biologicals
NB10066413 |
0.5 mL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
NB017771
|
Novus Biologicals
NB100664130.1ML |
0.1 mL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
Citidine Deaminase Polyclonal antibody specifically detects Citidine Deaminase in Human, Yeast samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Citidine Deaminase | |
Western Blot 1:100-1:500, ELISA 1:500-1:2500, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
Polyclonal | |
Purified | |
RUO | |
Human, Yeast | |
Q12178 | |
978 | |
Purified yeast cytosine deaminase | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
5.0 mg/mL | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Sheep | |
Lipid and Metabolism | |
PBS with 0.09% Sodium Azide | |
CDDCytidine aminohydrolase, cytidine deaminase, cytosine nucleoside deaminase, EC 3.5.4.5, small cytidine deaminase | |
CDA | |
IgG | |
Protein G purified | |
This is specific for cytosine deaminase. It recognises the native antigen in ELISA, and in Western blot two bands were visualised by ECL at 13kD and 17kD. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title