Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Citraconic hydrazide, 97%

Click to view available options
Quantity:
1g
5g
25g
5754-18-7
C5H6N2O2
126.115
MFCD00067746
QAVYOWFNXMHVEL-UHFFFAOYSA-N
4-methylpyridazine-3,6-diol,3,6-dihydroxy-4-methylpyridazine,1,2-dihydro-4-methyl-3,6-pyridazinedione,4-methyl-3,6-pyridazinediol,3,6-pyridazinedione, 1,2-dihydro-4-methyl,3, 1,2-dihydro-4-methyl,2-butenedioic acid, cyclic hydrazide, z,acmc-1aqja,4-methylmaleic hydrazide,methylmaleic acid hydrazide
79826
4-methyl-1,2-dihydropyridazine-3,6-dione
CC1=CC(=O)NNC1=O
5754-18-7
C5H6N2O2
126.115
MFCD00067746
QAVYOWFNXMHVEL-UHFFFAOYSA-N
4-methylpyridazine-3,6-diol,3,6-dihydroxy-4-methylpyridazine,1,2-dihydro-4-methyl-3,6-pyridazinedione,4-methyl-3,6-pyridazinediol,3,6-pyridazinedione, 1,2-dihydro-4-methyl,3, 1,2-dihydro-4-methyl,2-butenedioic acid, cyclic hydrazide, z,acmc-1aqja,4-methylmaleic hydrazide,methylmaleic acid hydrazide
79826
4-methyl-1,2-dihydropyridazine-3,6-dione
CC1=CC(=O)NNC1=O
Melting Point 284°C to 288°C
Quantity 1g
Formula Weight 126.11
Percent Purity 97%
Chemical Name or Material Citraconic hydrazide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning