Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Citrate Buffer, 0.2M buffer soln., pH 5.0, low endotoxin

Click to view available options
Quantity:
100mL
250mL
6132-04-3
C6H9Na3O9
294.10
MFCD00150031
NLJMYIDDQXHKNR-UHFFFAOYSA-K
71474
CHEBI:32142
trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]
6132-04-3
C6H9Na3O9
294.10
MFCD00150031
NLJMYIDDQXHKNR-UHFFFAOYSA-K
71474
CHEBI:32142
trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]
Concentration 0.2M buffer soln.
Physical Form Liquid
Odor Odorless
pH 5.0
Color Colorless
Quantity 100mL
Chemical Name or Material Citrate Buffer, low endotoxin

GHS H Statement
H319
Causes serious eye irritation.

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

TSCA : TSCA

storageNote1 : 2° to 8°C