Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Citric acid, Anhydrous, 99.5 to 100.5% (Dry Basis), Thermo Scientific™

Click to view available options
Quantity:
500g
1kg
Packaging:
Plastic bottle
77-92-9
C6H8O7
192.123
MFCD00011669
KRKNYBCHXYNGOX-UHFFFAOYSA-N
citric acid,citric acid, anhydrous,citro,anhydrous citric acid,citrate,aciletten,citretten,chemfill,hydrocerol a,1,2,3-propanetricarboxylic acid, 2-hydroxy
311
CHEBI:30769
2-hydroxypropane-1,2,3-tricarboxylic acid
C(C(=O)O)C(CC(=O)O)(C(=O)O)O
77-92-9
C6H8O7
192.123
MFCD00011669
KRKNYBCHXYNGOX-UHFFFAOYSA-N
citric acid,citric acid, anhydrous,citro,anhydrous citric acid,citrate,aciletten,citretten,chemfill,hydrocerol a,1,2,3-propanetricarboxylic acid, 2-hydroxy
311
CHEBI:30769
2-hydroxypropane-1,2,3-tricarboxylic acid
C(C(=O)O)C(CC(=O)O)(C(=O)O)O
Formula Weight 192.12
Physical Form Crystalline Powder
Packaging Plastic bottle
Specific Gravity 1.665
Melting Point 153°C to 154°C
Quantity 500g
Chemical Name or Material Citric Acid Anhydrous