Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Citric acid, anhydrous, ACS, >99.5%, Alfa Aesar™
Click to view available options
Quantity:
100g
500g
2kg
Specifications
Assay Percent Range | >99.5+% |
Beilstein | 782061 |
Solubility Information | It is very soluble in water,freely soluble in ethanol (96 %) and sparingly soluble in ether. |
Density | 1.665 |
Formula Weight | 192.13 |
Physical Form | Crystalline |
Grade | ACS Reagent |
Melting Point | 153°C to 154°C |
Quantity | 500g |
Sensitivity | Hygroscopic |
Show More |
Safety and Handling
P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c
H319-H335-H500
EINECSNumber : 201-069-1
RTECSNumber : GE7350000
TSCA : Yes
Recommended Storage : Ambient temperatures