Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citric Acid, Anhydrous, Powder, FCC, 99.5-100.5%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
77-92-9
C6H8O7
192.12
KRKNYBCHXYNGOX-UHFFFAOYSA-N
2-hydroxypropane-1,2,3-tricarboxylic acid
OC(=O)CC(O)(CC(O)=O)C(O)=O
77-92-9
C6H8O7
192.12
KRKNYBCHXYNGOX-UHFFFAOYSA-N
2-hydroxypropane-1,2,3-tricarboxylic acid
OC(=O)CC(O)(CC(O)=O)C(O)=O

C1282, 77-92-9, C6H8O7

Spectrum™ Citric Acid, Anhydrous, Powder, FCC is the most widely used organic acid and pH-control agent in foods and beverages. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Ignition Residue 0.05%
Percent Purity 99.5-100.5%
Lead (Pb) 0.5 mg/kg
Packaging Poly Pail
Water 0.5%
Quantity 2.5 kg