Learn More
Citric Acid, Anhydrous, Powder, FCC, 99.5-100.5%, Spectrum™
Click to view available options
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Quantity:
500 g
C1282, 77-92-9, C6H8O7
Spectrum™ Citric Acid, Anhydrous, Powder, FCC is the most widely used organic acid and pH-control agent in foods and beverages. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Ignition Residue 0.0005
Lead (Pb) 0.5 mg/kg
Packaging Poly Pail
Water 0.005