Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citric acid, >=99.5%, ACS reagent, anhydrous, Thermo Scientific Chemicals

Catalog Number ac423560100
Click to view available options
Quantity:
10 kg
Packaging:
Plastic drum
77-92-9
C6H8O7
192.12
MFCD00011669
KRKNYBCHXYNGOX-UHFFFAOYSA-N
citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy
311
CHEBI:30769
2-hydroxypropane-1,2,3-tricarboxylic acid
OC(=O)CC(O)(CC(O)=O)C(O)=O
77-92-9
C6H8O7
192.12
MFCD00011669
KRKNYBCHXYNGOX-UHFFFAOYSA-N
citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy
311
CHEBI:30769
2-hydroxypropane-1,2,3-tricarboxylic acid
OC(=O)CC(O)(CC(O)=O)C(O)=O

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 153.0°C
Color White
Assay Percent Range 99.6%
Linear Formula HO2CCH2C(OH)(CO2H)CH2CO2H
Solubility Information Solubility in water: 750g/L (20°C). Other solubilities: soluble in alcohol and ether, badly soluble in diethyl ether, insoluble in benzene and chloroform
Formula Weight 192.13
Percent Purity 99.6%
Grade ACS Reagent
Quantity 10 kg
Flash Point 100°C
Infrared Spectrum Authentic
Packaging Plastic drum
Physical Form Crystalline Powder
Residue after Ignition 0.02% max.
Chemical Name or Material Citric acid
Show More Show Less
Hazard Category Warning

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit