Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citric Acid, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle
5949-29-1
C6H10O8
210.138
YASYEJJMZJALEJ-UHFFFAOYSA-N
citric acid monohydrate,citric acid hydrate,2-hydroxypropane-1,2,3-tricarboxylic acid hydrate,citric acid, monohydrate,unii-2968phw8qp,1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate,citrate,acidum citricum monohydricum,citric acid monohydrate usp,citric acid monohydrate usp
22230
CHEBI:31404
2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O
5949-29-1
C6H10O8
210.138
YASYEJJMZJALEJ-UHFFFAOYSA-N
citric acid monohydrate,citric acid hydrate,2-hydroxypropane-1,2,3-tricarboxylic acid hydrate,citric acid, monohydrate,unii-2968phw8qp,1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate,citrate,acidum citricum monohydricum,citric acid monohydrate usp,citric acid monohydrate usp
22230
CHEBI:31404
2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O

High quality chemicals for laboratory and specialized industrial use

Formula Weight 210.14
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Quantity 500g
Chemical Name or Material Citric Acid