Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citric acid, diammonium salt, ACS reagent, Acros Organics

Click to view available options
Quantity:
100g
500g
2.5kg
3012-65-5
C6H14N2O7
226.19
YXVFQADLFFNVDS-UHFFFAOYSA-N
ammonium citrate dibasic,ammonium hydrogencitrate,diammonium hydrogen citrate,citric acid diammonium salt,citric acid ammonium salt,di-ammonium hydrogen citrate,citro diamine,citric acid diamine,ammonium citrate, dibasic, acs,ammonium citrate dibasic, puriss
13710713
CHEBI:63076
azane;2-hydroxypropane-1,2,3-tricarboxylic acid
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.N
3012-65-5
C6H14N2O7
226.19
YXVFQADLFFNVDS-UHFFFAOYSA-N
ammonium citrate dibasic,ammonium hydrogencitrate,diammonium hydrogen citrate,citric acid diammonium salt,citric acid ammonium salt,di-ammonium hydrogen citrate,citro diamine,citric acid diamine,ammonium citrate, dibasic, acs,ammonium citrate dibasic, puriss
13710713
CHEBI:63076
azane;2-hydroxypropane-1,2,3-tricarboxylic acid
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.N
Phosphate 0.0005% max
Chloride 0.001% max
Residue after Ignition 0.01% max
Insoluble Matter 0.005% max
Quantity 100g
Density 1.5000g/mL
Specific Gravity 1.5
Merck Index 15,509
Solubility Information Solubility in water: soluble
Formula Weight 226.19
Grade ACS Reagent
Chemical Name or Material Citric acid, diammonium salt
Show More Show Less

GHS Signal Word: Warning

EINECSNumber : 221-146-3