Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citric Acid, Granular, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle

Chemical Identifiers

5949-29-1
C6H10O8
210.138
YASYEJJMZJALEJ-UHFFFAOYSA-N
22230
CHEBI:31404
2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O

High quality chemicals for laboratory and specialized industrial use

Formula Weight 210.14
Physical Form Granular
Packaging Poly Bottle
Specific Gravity 1L = 1.54kg
Quantity 2.5kg
Chemical Name or Material Citric Acid
Grade Baker Analyzed™ ACS Reagent