Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citric acid monohydrate, 99.5%, for analysis, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
2.5kg
Packaging:
Plastic bottle
5949-29-1
C6H10O8
210.138
MFCD00149972
YASYEJJMZJALEJ-UHFFFAOYSA-N
citric acid monohydrate,citric acid hydrate,2-hydroxypropane-1,2,3-tricarboxylic acid hydrate,citric acid, monohydrate,unii-2968phw8qp,1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate,citrate,acidum citricum monohydricum,citric acid monohydrate usp
22230
CHEBI:31404
2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O
5949-29-1
C6H10O8
210.138
MFCD00149972
YASYEJJMZJALEJ-UHFFFAOYSA-N
citric acid monohydrate,citric acid hydrate,2-hydroxypropane-1,2,3-tricarboxylic acid hydrate,citric acid, monohydrate,unii-2968phw8qp,1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate,citrate,acidum citricum monohydricum,citric acid monohydrate usp
22230
CHEBI:31404
2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O
Biological buffer for analysis
C2O4 500ppm max.
Assay Percent Range 99.5%
Linear Formula HO2CCH2C(OH)(CO2H)CH2CO2H·H2O
Chlorine (Cl) 10ppm max.
Merck Index 15, 2325
Solubility Information Solubility in water: 676g/L (25°C). Other solubilities: 2.17g/100g ether - 0.007g/100g chloroform, 15.43g/100g amyl alcohol - 5.98g/100g amyl acetate, 5.28g/100g ethyl acetate - 197g/100g methanol, 62.8g/100g propanol
Density 1.54g/mL
Formula Weight 210.15
Physical Form Crystals or Crystalline Powder
Percent Purity 99.5%
Grade Analysis
Heavy Metals (as Pb) 5ppm max.
Infrared Spectrum Authentic
Iron (Fe) 3ppm max.
Packaging Plastic bottle
Readily Carbonizable Subtances Passes Test
Solubility Solubility in water: 676g/L (25°C). Other solubilities: 2.17g/100g ether - 0.007g/100g chloroform, 15.43g/100g amyl alcohol - 5.98g/100g amyl acetate, 5.28g/100g ethyl acetate - 197g/100g methanol, 62.8g/100g propanol
Sulfate 50ppm max.
Sulfated Ash 0.1% max.
Water 7.5 to 9% (K.F.)
Melting Point 135.0°C to 152.0°C
Color White
Quantity 2.5kg
Flash Point 174°C
Chemical Name or Material Citric acid monohydrate
Show More Show Less

GHS H Statement:
Causes serious eye irritation.

GHS P Statement:
Wash face, hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning