Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citric acid monohydrate, 99+%, ACS reagent, Acros Organics

Click to view available options
Quantity:
250g
1kg
2.5kg
Packaging:
Plastic bottle
5949-29-1
C6H8O7·H2O
210.15
YASYEJJMZJALEJ-UHFFFAOYSA-N
citric acid monohydrate,citric acid hydrate,2-hydroxypropane-1,2,3-tricarboxylic acid hydrate,citric acid, monohydrate,unii-2968phw8qp,1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate,citrate,acidum citricum monohydricum,citric acid monohydrate usp
22230
CHEBI:31404
2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O
5949-29-1
C6H8O7·H2O
210.15
YASYEJJMZJALEJ-UHFFFAOYSA-N
citric acid monohydrate,citric acid hydrate,2-hydroxypropane-1,2,3-tricarboxylic acid hydrate,citric acid, monohydrate,unii-2968phw8qp,1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate,citrate,acidum citricum monohydricum,citric acid monohydrate usp
22230
CHEBI:31404
2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O
Chloride 0.001% max
Phosphate 0.001% max
Insoluble Matter 0.005% max
Sulfate 0.002% max
Residue after Ignition 0.02% max
Flash Point 174°C
Melting Point 135.0°C to 152.0°C
Quantity 1kg
Packaging Plastic bottle
Assay Percent Range 99+%
Linear Formula HO2CCH2C(OH)(CO2H)CH2CO2H·H2O
Merck Index 15,2325
Solubility Information Solubility in water: 676g/L (25°C). Other solubilities: 2.17g/100g ether - 0.007g/100g chloroform, 15.43g/100g amyl alcohol - 5.98g/100g amyl acetate, 5.28g/100g ethyl acetate - 197g/100g methanol, 62.8g/100g propanol
Formula Weight 210.15
Percent Purity 99+%
Grade ACS Reagent
Chemical Name or Material Citric acid monohydrate
Show More Show Less

GHS H Statement
Causes serious eye irritation.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact len

GHS Signal Word: Warning