Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citric Acid Monohydrate, ACS Reagent, Reagents

Catalog No. 01338182
Change view
Click to view available options
Quantity:
500 g
3 kg
10 kg
25 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-338-182 25 kg
01-338-183 10 kg
01-338-181 3 kg
01-338-180 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-338-182 Supplier Reagents Holdings Llc Supplier No. C110450025F4
Only null left
Add to Cart
Add to Cart

Citric Acid Monohydrate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 5949-29-1
Molecular Formula C6H10O8
Molecular Weight (g/mol) 210.14
InChI Key YASYEJJMZJALEJ-UHFFFAOYSA-N
Synonym 2-hydroxypropane-1,2,3-tricarboxylic Acid monohydrate
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate
SMILES O.OC(=O)CC(O)(CC(O)=O)C(O)=O

Specifications

Melting Point 153°C
Color Colorless/White
Grade ACS
Quantity 25 kg
Packaging Fiber Drum
Physical Form Solid
Specific Gravity 1.542
Chemical Name or Material Citric Acid Monohydrate, ACS Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.