Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citric Acid, Monohydrate, Crystal, ACS, 99.0-102.0%, Spectrum™

Click to view available options
Quantity:
25 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5949-29-1
5949-29-1
CAS Max % 1
Chloride 1e-05
Percent Purity 99 to 102%
Grade ACS
Insoluble Matter 5e-05
Iron (Fe) 3 ppm
Lead (Pb) 2 ppm
Packaging Poly Bottle
Phosphate 1e-05
Residue after Ignition 0.0002
Quantity 2.5 kg
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.