Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citric Acid, Monohydrate, Granular, FCC, 99.5-100.5%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5949-29-1
C6H10O8
210.14
YASYEJJMZJALEJ-UHFFFAOYSA-N
2-hydroxypropane-1,2,3-tricarboxylic acid hydrate
O.OC(=O)CC(O)(CC(O)=O)C(O)=O
5949-29-1
C6H10O8
210.14
YASYEJJMZJALEJ-UHFFFAOYSA-N
2-hydroxypropane-1,2,3-tricarboxylic acid hydrate
O.OC(=O)CC(O)(CC(O)=O)C(O)=O

C1283, 5949-29-1, C6H8O7•H2O

Spectrum™ Citric Acid, Monohydrate, Granular, FCC is widely used as acidity regulator and presevative in food production and as an acidity regulator and antioxidant in beverages. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Ignition Residue 0.05%
Percent Purity 99.5-100.5%
Lead (Pb) 0.5 mg/kg
Packaging Poly Pail
Water 8.8%
Quantity 2.5 kg