Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citric Acid Monohydrate (Granular/USP), Fisher Chemical

Click to view available options
Quantity:
3kg
10kg
50kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5949-29-1
C6H10O8
210.138
MFCD00149972
YASYEJJMZJALEJ-UHFFFAOYSA-N
citric acid monohydrate,citric acid hydrate,2-hydroxypropane-1,2,3-tricarboxylic acid hydrate,citric acid, monohydrate,unii-2968phw8qp,1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate,citrate,acidum citricum monohydricum,citric acid monohydrate usp
22230
CHEBI:31404
2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O
5949-29-1
C6H10O8
210.138
MFCD00149972
YASYEJJMZJALEJ-UHFFFAOYSA-N
citric acid monohydrate,citric acid hydrate,2-hydroxypropane-1,2,3-tricarboxylic acid hydrate,citric acid, monohydrate,unii-2968phw8qp,1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate,citrate,acidum citricum monohydricum,citric acid monohydrate usp
22230
CHEBI:31404
2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O
Clarity Pass Test (USP)
Formula Weight 210.14
Endotoxin Report EU/mg (USP)
Physical Form Solid
Percent Purity 99.5 to 100.5% (USP)
Grade USP
Heavy Metals (as Pb) 0.001% max. (USP)
Identification Pass Test (USP)
Ignition Residue 0.1% max. (USP)
Oxalic Acid 0.036% max. (USP)
Packaging Fiber Drum
Readily Carbonizable Subtances Pass Test (USP)
Residual Solvents Meets Requirements (USP)
Sulfate 0.015% max. (USP)
Water 7.5 to 9.0% (USP)
Melting Point 135°C
pH 2.2
Color White
Quantity 50kg
Chemical Name or Material Citric Acid Monohydrate
Show More Show Less

WARNING!

Emergency Overview
Risk of serious damage to eyes. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:2
Flammability:1
Instability:0