Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Citric acid, monosodium salt, 99%, pure, anhydrous, ACROS Organics™
Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass bottle
Plastic bottle
Specifications
Quantity | 500g |
Packaging | Glass bottle |
Infrared Spectrum | Authentic |
Color | White |
Melting Point | 209.0°C to 215.0°C |
Assay Percent Range | 99% |
Linear Formula | HO2CCH2C(OH)(CO2H)CH2CO2Na |
Beilstein | 03, 563 |
Solubility Information | Solubility in water: freely soluble |
Formula Weight | 214.11 |
Show More |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Cont
GHS Signal Word: Warning
EINECSNumber : 242-734-6
RTECSNumber : GE7950000
TSCA : TSCA