Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citric Acid, Powder, U.S.P., J.T.Baker™

Click to view available options
Quantity:
500g
2kg
Packaging:
Poly Bottle
77-92-9
C6H8O7
192.123
KRKNYBCHXYNGOX-UHFFFAOYSA-N
citric acid,citric acid, anhydrous,citro,anhydrous citric acid,citrate,aciletten,citretten,chemfill,hydrocerol a,1,2,3-propanetricarboxylic acid, 2-hydroxy
311
CHEBI:30769
2-hydroxypropane-1,2,3-tricarboxylic acid
C(C(=O)O)C(CC(=O)O)(C(=O)O)O
77-92-9
C6H8O7
192.123
KRKNYBCHXYNGOX-UHFFFAOYSA-N
citric acid,citric acid, anhydrous,citro,anhydrous citric acid,citrate,aciletten,citretten,chemfill,hydrocerol a,1,2,3-propanetricarboxylic acid, 2-hydroxy
311
CHEBI:30769
2-hydroxypropane-1,2,3-tricarboxylic acid
C(C(=O)O)C(CC(=O)O)(C(=O)O)O
Formula Weight 192.12
Physical Form Powder
Grade USP
Packaging Poly Bottle
Specific Gravity 1L = 1.44kg
Quantity 500g
Chemical Name or Material Citric Acid