Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citric Acid, Powder, U.S.P., J.T. Baker™

Catalog Number p-5391483
Click to view available options
Quantity:
500 g
2 kg
Packaging:
Poly Bottle
77-92-9
C6H8O7
192.12
MFCD00011669
KRKNYBCHXYNGOX-UHFFFAOYSA-N
citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy
311
CHEBI:30769
2-hydroxypropane-1,2,3-tricarboxylic acid
OC(=O)CC(O)(CC(O)=O)C(O)=O
77-92-9
C6H8O7
192.12
MFCD00011669
KRKNYBCHXYNGOX-UHFFFAOYSA-N
citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy
311
CHEBI:30769
2-hydroxypropane-1,2,3-tricarboxylic acid
OC(=O)CC(O)(CC(O)=O)C(O)=O
Formula Weight 192.12
Grade USP
Quantity 500 g
Packaging Poly Bottle
Physical Form Powder
Specific Gravity 1L = 1.44kg
Chemical Name or Material Citric Acid
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit