Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Citric Acid, Powder, U.S.P., J.T.Baker™
Click to view available options
Quantity:
500g
2kg
Packaging:
Poly Bottle
Specifications
Formula Weight | 192.12 |
Physical Form | Powder |
Grade | USP |
Packaging | Poly Bottle |
Specific Gravity | 1L = 1.44kg |
Quantity | 2kg |
Chemical Name or Material | Citric Acid |