Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citric acid, trilithium salt tetrahydrate, 98%, pure, ACROS Organics™

Click to view available options
Quantity:
5g
500g
Packaging:
Glass bottle
Plastic bottle
6080-58-6
C6H5Li3O7·4H2O
281.97
MFCD00150030
HXGWMCJZLNWEBC-UHFFFAOYSA-K
lithium citrate tetrahydrate,unii-5z6e9k79yv,trilithium citrate tetrahydrate,lithium citrate usp,citric acid trilithium salt tetrahydrate,tri-lithium citrate tetrahydrate,lithium citrate tribasic tetrahydrate,lithonate tn,1,2,3-propanetricarboxylic acid, 2-hydroxy-trilithium salt tetrahydrate
2724118
CHEBI:64754
trilithium;2-hydroxypropane-1,2,3-tricarboxylate;tetrahydrate
[Li+].[Li+].[Li+].C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.O.O
6080-58-6
C6H5Li3O7·4H2O
281.97
MFCD00150030
HXGWMCJZLNWEBC-UHFFFAOYSA-K
lithium citrate tetrahydrate,unii-5z6e9k79yv,trilithium citrate tetrahydrate,lithium citrate usp,citric acid trilithium salt tetrahydrate,tri-lithium citrate tetrahydrate,lithium citrate tribasic tetrahydrate,lithonate tn,1,2,3-propanetricarboxylic acid, 2-hydroxy-trilithium salt tetrahydrate
2724118
CHEBI:64754
trilithium;2-hydroxypropane-1,2,3-tricarboxylate;tetrahydrate
[Li+].[Li+].[Li+].C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.O.O
Quantity 500g
Melting Point 112.0°C
Packaging Plastic bottle
Color White
Water 24% max.
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula LiO2CCH2C(OH)(CO2Li)CH2CO2Li·4H2O
Beilstein 03, I, 196
Merck Index 15, 5586
Solubility Information Solubility in water: 745g/L water (25°C). Other solubilities: slightly soluble in alcohol
Formula Weight 281.97
Physical Form Fine Crystalline Powder
Percent Purity 98%
Grade Pure
Chemical Name or Material Citric acid, trilithium salt tetrahydrate, 98%
Show More Show Less