Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citric Acid, U.S.P. - F.C.C., J.T.Baker™

Click to view available options
Quantity:
2.5kg
500g
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle

Chemical Identifiers

5949-29-1
C6H10O8
210.138
YASYEJJMZJALEJ-UHFFFAOYSA-N
22230
CHEBI:31404
CAS 5949-29-1
Molecular Formula C6H10O8
InChI Key YASYEJJMZJALEJ-UHFFFAOYSA-N
SMILES C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate
Molecular Weight (g/mol) 210.138
PubChem CID 22230
ChEBI CHEBI:31404
Formula Weight 210.14
Packaging Poly Bottle
Specific Gravity 1L = 1.54kg
Quantity 2.5kg
Chemical Name or Material Citric Acid
Grade FCC/USP
Show More Show Less