Available on GSA/VA Contract for Federal Government customers only. Citrisolv Clearing Agent, Fisherbrand

$270.01

Chemical Identifiers

Chemical Name or Material Fisherbrand™ CitriSolv™ Clearing Agent
CAS [5989-27-5, 97.00-98.00%], [25013-16-5, 0.20-0.24%]

  • Products 1
  • Description & Specifications
Products
Catalog Number Description Price Quantity Availability  

22-143-975

Available on GSA/VA Contract for Federal Government customers only. 
Clearing agent; 1 gal.
Each for $270.01
Case of 4 EA for $548.05
Check Availability
Add to Cart
Descriptions & Specifications
Safety and Handling

Related Products