Learn More
Citrulline Mouse anti-Non-species specific, HRP, Clone: 2D3.1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259364H
Catalog No. NBP259364H
Add to cart
Description
Description
Citrulline Monoclonal antibody specifically detects Citrulline in Non-species specific samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
Citrulline
2D3.1
Western Blot, ELISA
Purified
Synthetic L-Citrulline conjugated to Keyhole Limpet Hemocyanin (KLH).
Protein G purified
Store at 4C in the dark.
Monoclonal
Non-species specific
Western Blot, ELISA
HRP
PBS with No Preservative
Mouse
IgG1
100 ul
Primary
Specific for citrulline residues on modified proteins. Tested using citrulline conjugated Bovine Serum Albumin (BSA).