Learn More
Aviva Systems Biology Corp CIZ1 ANTIBODY 50UG/50UL DFS Item
Manufacturer:  Aviva Systems Biology Corp ARP38956_P050
This is a rabbit polyclonal antibody against CIZ1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human CIZ1. Alternate names: LSFR1, ZNF356, NP94ApplicationIHC, WB. Species reactivity: Horse, Human
Catalog No. 50-014-53121