Learn More
Tocris Bioscience CK 869
Arp2/3 inhibitor; inhibits actin polymerization
Manufacturer: Tocris Bioscience 4984/50
This item is not returnable.
View return policy
Description
Inhibitor of human and bovine actin-related protein 2/3 (Arp2/3) complex. Inhibits actin polymerization (IC50= 11 μM). Does not inhibit budding or fission yeast Arp2/3 complexes.Specifications
CK 869 | |
>98 | |
50mg |
CK 869 | |
394.28 |