Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris™ CK 869

Arp2/3 inhibitor; inhibits actin polymerization

Manufacturer:  Tocris™ 4984/10

 View more versions of this product

Catalog No. 49-841-0R


  Check Availability
Add to cart

Disclaimers:

For Research Use Only


Specifications

Description

Inhibitor of human and bovine actin-related protein 2/3 (Arp2/3) complex. Inhibits actin polymerization (IC50= 11µM). Does not inhibit budding or fission yeast Arp2/3 complexes.
  • Chemical Name: 2-(3-Bromophenyl)-3-(2,4-dimethoxyphenyl)-4-thiazolidinone
  • Formula: C17H16BrNO3S
  • CAS Number: 388592-44-7
  • Solubility: Soluble to 100mM in DMSO
  • Purity: >98
Description & Specifications

Specifications

CK 869
>98
10mg
CK 869
394.28
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit