Learn More
Pointe Scientific CK Reagents
Supplier:  Pointe Scientific HC922250
 View more versions of this product
Catalog No. 23-666-213
Add to cart
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
CE Marked
Creatine Kinase Reagent Kit
Quantitative determination of Creatine Kinase activity in Serum
2 x 100mL R1, 2 x 25mL R2
Liquid
CK R1 (Buffer) contains: Imidazole Buffer 100mM/L (pH 6.7), NADP 2mM/L, HK (Baker's Yeast) 2.5KU/L, Glucose 20mM/L, Magnesium Acetate 10mM/L, EDTA 2mM/L, N-acetylcysteine (NAC) 20mM/L, CK R2 (enzyme reagent) contains: Imidazole Buffer 100mM/L (pH 6.7), A
Hitachi 717 analyzers