Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CK1 gamma-1 Monoclonal Antibody (OTI1C5), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA526787
Description
CK1 gamma-1 Monoclonal antibody specifically detects CK1 gamma-1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
CK1 gamma-1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9HCP0 | |
Csnk1g1 | |
Human recombinant protein fragment corresponding to amino acids 163-422 of CSNK1G1 produced in E.coli | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1C5 | |
Unconjugated | |
Csnk1g1 | |
9130020E21Rik; casein kinase 1 gamma 1; casein kinase 1, gamma 1; casein kinase I isoform gamma-1; casein kinase I, gamma 1; CK I gamma1; CK1G1; CK1gamma1; CKI gamma 1; CKI gamma1; CK-I gamma1; CKI- gamma1; CKI gamma-1; CKI-gamma 1; CKI-gamma-1; CSNK1G1; KC1G1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
53944 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction