Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CK1 gamma-1 Mouse anti-Human, Clone: OTI1D7, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526789

Catalog No. PIMA526789


Add to cart

Description

Description

Casein kinase I (also designated CKI) and casein kinase II (also designated CKII) compose a family of serine/ threonine protein kinases which are present in all eukaryotes examined to date. CKI family members, which include CKI alpha, gamma, epsilon and delta, have been implicated in the control of cytoplasmic and nuclear processes, including DNA replication and repair. CKII is usually expressed as a tetrameric complex consisting of either an alpha2 beta2 or an alpha alpha ft. beta2 structure. The alpha catalytic subunit is stimulated by the beta regulatory subunit, which undergoes autophosphorylation. CKII activity is high in the cytosol and nucleus of proliferating and differentiating cells. CKII is known to phosphorylate more than 100 different substrates including nuclear oncoproteins, transcription factors and enzymes involved in DNA metabolism.
Specifications

Specifications

CK1 gamma-1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9HCP0
Casein kinase I isoform gamma-1, CKI-gamma 1, CSNK1G1, KC1G1, CK1G1, CKI gamma1, CK-I gamma1, CKI- gamma1, CKI gamma-1, CKI gamma1, CK I gamma1, CKI gamma1, CKI gamma 1, CKI gamma 1, CKI-gamma-1
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
53944
Immunohistochemistry (Paraffin), Western Blot
OTI1D7
Unconjugated
CSNK1G1
Liquid
CSNK1G1
Human recombit protein fragment corresponding to amino acids 163-422 of CSNK1G1 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit