Learn More
CKAP1 Rabbit anti-Human, Alexa Fluor 647, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP300264AF647
Catalog No. NB065176
Add to cart
Description
Description
CKAP1 Polyclonal specifically detects CKAP1 in Human samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
CKAP1
Alexa Fluor 647
50mM Sodium Borate
CG22Tubulin-specific chaperone B, CKAP1MGC14625, CKAPI, cytoskeleton associated protein 1, Cytoskeleton-associated protein 1Cytoskeleton-associated protein CKAPI, tubulin folding cofactor B, tubulin-folding cofactor B
Produced in rabbits immunized with purified, recombinant CKAP1 (Q99426; Met1-Ile244).
Affinity purified
RUO
Primary
1155
Western Blot, ELISA
Western Blot, ELISA
Purified
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Polyclonal
Human