Learn More
CKMT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA549789
Catalog No. PA549789
Add to cart
Description
Description
The antibody detects endogenous levels of total CKMT2 protein.
Mitochondrial creatine kinase (MtCK) is responsible for the transfer of high energy phosphate from mitochondria to the cytosolic carrier, creatine. It belongs to the creatine kinase isoenzyme family. It exists as two isoenzymes, sarcomeric MtCK and ubiquitous MtCK, encoded by separate genes. Mitochondrial creatine kinase occurs in two different oligomeric forms: dimers and octamers, in contrast to the exclusively dimeric cytosolic creatine kinase isoenzymes. Sarcomeric mitochondrial creatine kinase has 80% homology with the coding exons of ubiquitous mitochondrial creatine kinase. This gene contains sequences homologous to several motifs that are shared among some nuclear genes encoding mitochondrial proteins and thus may be essential for the coordinated activation of these genes during mitochondrial biogenesis. Three transcript variants encoding the same protein have been found for this gene.
Specifications
Specifications
CKMT2
Polyclonal
Unconjugated
CKMT2
Liquid
CKMT2
Synthesized peptide derived from internal of human CKMT2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P17540, Q6P8J7, P09605
Creatine kinase, sarcomeric mitochondrial [Precursor], EC 2.7.3.2, S-MtCK, Mib-CK
Rabbit
IgG
100 μL
-20°C
Primary
1160, 688698, 76722