Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CKS2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA592063
Description
Immunogen sequence: MAHKQIYYSD KYFDEHYEYR HVMLPRELSK QVPKTHLMSE EEWRRLGVQQ SLGWVHYMIH EPEPHILLFR RPLPKDQQK; Positive Samples: Rat liver.
CKS2 protein binds to the catalytic subunit of the cyclin dependent kinases and is essential for their biological function. The CKS2 mRNA is found to be expressed in different patterns through the cell cycle in HeLa cells, which reflects specialized role for the encoded protein. [provided by RefSeq, Jul 2008].
Specifications
CKS2 | |
Polyclonal | |
Unconjugated | |
Cks2 | |
1110038L14Rik; CDC28 protein kinase 2; CDC28 protein kinase regulatory subunit 2; CKS1(S. cerevisiae Cdc28/Cdc2 kinase subunit) homolog-2; CKS2; CKS-2; CKSHS2; Cyclin-dependent kinases regulatory subunit 2; RGD1562047; RP11-8C21.2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
498709 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P56390 | |
Cks2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-79 of human CKS2 (NP_0018181). | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction