Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cl-4AS-1, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent steroidal androgen receptor agonist (IC50 = 12 nM). Mimics the action of 5α-dihydrotestosterone (DHT). Transactivates the mouse mammary tumor virus (MMTV) promoter; represses MMP1 promoter activity. Inhibits 5α-reductase type I and II (IC50 values are 6 and 10 nM respectively).
Specifications
Quantity | 10 mg |
Formula Weight | 441.01 |
Percent Purity | >99% |
Chemical Name or Material | Cl-4AS-1 |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction