Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CL 82198 hydrochloride, Tocris Bioscience™

Catalog Number p-4783867
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
1188890-36-9
C17H23ClN2O3
338.832
PIOACXKZWXHBRB-UHFFFAOYSA-N
cl 82198 hydrochloride, n-4-morpholinobutyl benzofuran-2-carboxamide hydrochloride, n-4-4-morpholinyl butyl-2-benzofurancarboxamide hydrochloride, cl-82198 calbiochem cat. no. 233105, n-4-4-morpholinyl butyl-1-benzofuran-2-carboxamide hydroc, n-4-morpholin-4-yl butyl-1-benzofuran-2-carboxamide hydrochloride, n-4-morpholin-4-yl butyl-1-benzofuran-2-carboxamide-hydrogen chloride 1/1
16760373
N-(4-morpholin-4-ylbutyl)-1-benzofuran-2-carboxamide;hydrochloride
C1COCCN1CCCCNC(=O)C2=CC3=CC=CC=C3O2.Cl
This item is not returnable. View return policy
1188890-36-9
C17H23ClN2O3
338.832
PIOACXKZWXHBRB-UHFFFAOYSA-N
cl 82198 hydrochloride, n-4-morpholinobutyl benzofuran-2-carboxamide hydrochloride, n-4-4-morpholinyl butyl-2-benzofurancarboxamide hydrochloride, cl-82198 calbiochem cat. no. 233105, n-4-4-morpholinyl butyl-1-benzofuran-2-carboxamide hydroc, n-4-morpholin-4-yl butyl-1-benzofuran-2-carboxamide hydrochloride, n-4-morpholin-4-yl butyl-1-benzofuran-2-carboxamide-hydrogen chloride 1/1
16760373
N-(4-morpholin-4-ylbutyl)-1-benzofuran-2-carboxamide;hydrochloride
C1COCCN1CCCCNC(=O)C2=CC3=CC=CC=C3O2.Cl

Selective inhibitor of MMP-13

Selective inhibitor of MMP-13 (89% inhibition at 10 μg/mL) that displays no activity at MMP-1, MMP-9 or TACE. Inhibits in vitro invasion by the human pituitary adenoma cell line HP75. Rescues paclitaxel induced axon degradation and reduces associated neurotoxicity in zebrafish.
Quantity 10 mg
Formula Weight 338.83
Percent Purity >98%
Chemical Name or Material CL 82198 hydrochloride
Recommended Storage Desiccate at Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit