Learn More
Cayman Chemical Cl-Amidine (hydrochloride), 1373232-26-8, 1 mg
SDP
Supplier:  Cayman Chemical 105991
Encompass_Preferred
A pan PAD inhibitor (IC50s = 0.8, 6.2, and 5.9 µM for PAD1, PAD3, and PAD4, respectively, in vitro); irreversibly inactivates PAD4 (kinact/KI = 13,000 M-1min-1) and PAD3 (kinact/KI = 2,000 M-1min-1); cytotoxic to HL-60, MCF7, and HT-29 cancer cell lines (IC50s = 0.25, 0.05 and 1 μM, respectively),
Catalog No. NC0194924
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.