Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cl-HIBO, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Subtype-selective and strongly desensitizing AMPA receptor agonist; selective for GluA1 and GluA2 subunit-containing receptors (EC50 values are 4.7, 1.7, 2700 and 1300 μM for rat recombinant homomeric GluA1-4 receptors respectively).
Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.
Specifications
Quantity | 50 mg |
Formula Weight | 206.59 |
Percent Purity | >99% |
Chemical Name or Material | Cl-HIBO |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction