Learn More
Selleck Chemical LLCSupplier Diversity Partner Cladribine 50mg 4291-63-8 2-CdA, 2-chlorodeoxyadenosine  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S119950MG
Cladribine (2-CdA, 2-chlorodeoxyadenosine) is an adenosine deaminase inhibitor for U266, RPMI8226, and MM1.S cells with IC50 of approximately 2.43 μM, 0.75 μM, and 0.18 μM, respectively.
Catalog No. 50-689-8